Learn More
Cayman Chemical UbIquItIn RemntK-E-GG Im 400uL
SDP
Catalog No. 502912455
Encompass_Preferred
An antiviral agent inhibits SARS-CoV-2 Mpro activity (IC50 0 081 UM) prevents HSV-1 and HSV-2 infection in Vero cells at 0 2 UM inhibits DDAH-1 and HDAC8 (IC50s 9 and 0 011 UM for the human enzymes respectively) inhibits bacterial KDO 8-P synthase (Ki 0 026 UM) reduces endothelial tube formation and LPS-induced NO production in primary human dermal microvascular endothelial cells
Catalog No. 50-291-2455 Supplier Cayman Chemical Supplier No. 34699400 UL
An antiviral agent inhibits SARS-CoV-2 Mpro activity (IC50 0 081 UM) prevents HSV-1 and HSV-2 infection in Vero cells at 0 2 UM inhibits DDAH-1 and HDAC8 (IC50s 9 and 0 011 UM for the human enzymes respectively) inhibits bacterial KDO 8-P synthase (Ki 0 026 UM) reduces endothelial tube formation and LPS-induced NO production in primary human dermal microvascular endothelial cells