Learn More
Enzo Life Sciences Ubiquitin-Rhodamine (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLSE7610025
Encompass_Preferred
Ubiquitin-Rhodamine is prepared by the C-terminal derivatization of ubiquitin with modified rhodamine-110 and has been shown to be a useful and sensitive fluorogenic substrate for wide range of deubiquitinylating enzymes (DUBs), including ubiquitin C-terminal hydrolases (UCHs) and ubiquitin specific proteases (USPs).  Ubiquitin-rhodamine is particularly useful for studying deubiquitinylating activity where detection sensitivity is essential.  The long excitation and emission wavelengths of rhodamine 110 (Excitation 485nm / Emission 520nm) avoid interference often caused at shorter wavelengths by small molecules and tissue culture components. The long excitation and emission wavelengths of rhodamine 110 (Excitation 485nm / Emission 520nm) avoid interference often caused at shorter wavelengths by small molecules and tissue culture components. Purity: ≥95% (HPLC). Formulation: Liquid. In dimethylsulfoxide. Source: Produced in E. coli and covalently attached to modified rhodamine-110.
Catalog No. 50-201-2100