Learn More
LifeSensors Ubiquitin-Rhodamine 50µg
SDP
Supplier:  LifeSensors SI02300050
Encompass_Preferred
Ubiquitin-Rhodamine 110 is a quenched, fluorescent substrate for deubiquitylases, especially ubiquitin C-terminal hydrolases. Cleavage of the amide bond between the C-terminal glycine of ubiquitin and rhodamine results in an increase in rhodamine fluorescence at 535 nm (485 nm excitation). Need bulk quantities? We would be happy to work out a custom price that fits your budget.
Catalog No. NC2285767