Learn More
Bioworld UBIQUITIN SEPAROPOREAGAROSE)
Catalog No. 50197766
Encompass
Ubiquitin-Separopore Covalently linked ubiquitin to Separopore beads via primary amine coupling leaving the C-terminus free. Useful for binding the ubiquitin activation enzyme (E1), ubiquitin carrier enzymes (E2s), ubiquitin ligases (E3...
Catalog No. 50-197-766 Supplier Bioworld Supplier No. 201810401
Ubiquitin-Separopore Covalently linked ubiquitin to Separopore beads via primary amine coupling leaving the C-terminus free. Useful for binding the ubiquitin activation enzyme (E1), ubiquitin carrier enzymes (E2s), ubiquitin ligases (E3...