Learn More
Bioworld UBIQUITIN SEPAROPOREAGAROSE)
Supplier:  Bioworld 201810402
Encompass
Ubiquitin-Separopore Covalently linked ubiquitin to Separopore beads via primary amine coupling leaving the C-terminus free. Useful for binding the ubiquitin activation enzyme (E1), ubiquitin carrier enzymes (E2s), ubiquitin ligases (E3...
Catalog No. 50-197-767