Learn More
Ubpbio Ubiquitin Vinyl Sulfone, 50UG
Supplier:  Ubpbio M1030
Encompass_Preferred
Ubiquitin Vinyl Sulfone (Ub-VS) is a potent and irreversible inhibitor of UCH, USP and OTU families of deubiquitinating enzymes (DUBs). It forms a covalent linkage with the active site Cys residue of a DUB. Ub-VS can be used to profile DUB and modify DUBs for structural determination.
Catalog No. 50-196-5174