Learn More
Enzo Life Sciences Ubiquitin vinyl methyl ester, (HA-tag) (25 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW08800025
Encompass_Preferred
Alternative name: HA-Ub-VME. MW: ~10’257Da. Purity: ≥95% (HPLC). Formulation: Lyophilized. Source: Synthetic. Use/Stability: Stable for at least 6 months after receipt when stored at -80°C. UniProt: P0CG47 (UBB), P0CG48 (UBC), P62979 (RPS27A), P62987 (UBA52). Handling: After reconstitution, prepare aliquots, store at -80°C and use within 1 month. Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC9986641