Learn More
Apexbio Technology LLC DiscoveryProbe™ Ubiquitination Compound Library, 100uL/well(10 mM solution),96 Well Racks With Screw Cap
SDP
Catalog No. 501998487
Encompass_Preferred
DiscoveryProbe™ Ubiquitination Compound Library is a collection of 144 ubiquitination compounds supplied as lyophilized powder or pre-dissolved DMSO solutions. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8487 Supplier Apexbio Technology LLC Supplier No. L1049.1
DiscoveryProbe™ Ubiquitination Compound Library is a collection of 144 ubiquitination compounds supplied as lyophilized powder or pre-dissolved DMSO solutions. Available in stock with overnight delivery and free shipping over $500 Cost-effective and competitive price to save your findings Potent, selective and cell-permeable in inhibiting or activating target molecules Diverse in chemical structure and route of administration (oral/i.m/i.v injection etc.) Detailed files describing potency, selectivity and applications etc. Supported by published data from top peer-reviewed journals Guaranteed high quality with NMR and HPLC validation. Other sizes are also available. Please inqury us for quote.