Learn More
Cayman Chemical UbIquItInlIkSpecIfIc Prote
SDP
Catalog No. 502945837
Encompass_Preferred
Source: Active recombinant S. cerevisiae N-terminal His-tagged Ulp1 expressed in E. coli • Amino acids: 403-621 • MW: 27.6 kDa
Catalog No. 50-294-5837 Supplier Cayman Chemical Supplier No. 41844500 UG
Source: Active recombinant S. cerevisiae N-terminal His-tagged Ulp1 expressed in E. coli • Amino acids: 403-621 • MW: 27.6 kDa