Learn More
LifeSensors UBITEST MAGNETIC TUBE ELUTION KIT
SDP
Catalog No. NC2285771
Encompass_Preferred
The UbiTest kit determines whether or not a target protein is poly-ubiquitinated. This powerful platform is an improvement on the most common method for investigating protein ubiquitination: immunoprecipitation and Western blot analysis. However, the substrate antibody often interacts differently with the poly-ubiquitinated forms of the substrate in the immuno-blotting step. This is due to epitope masking, reduced affinity, or changes in selectivity. A more definitive method for demonstrating protein ubiquitination is to couple immunoprecipitation with digestion by a broad spectrum deubiquitinase (DUB) prior to immunoblot analysis. An increased signal for the protein of interest (POI) after DUB treatment is a clear indication that the protein was ubiquitinated even if there was no clear reactivity in the untreated sample.
This item is not returnable. View return policy
Catalog No. NC2285771 Supplier LifeSensors Supplier No. UM0411M1000
The UbiTest kit determines whether or not a target protein is poly-ubiquitinated. This powerful platform is an improvement on the most common method for investigating protein ubiquitination: immunoprecipitation and Western blot analysis. However, the substrate antibody often interacts differently with the poly-ubiquitinated forms of the substrate in the immuno-blotting step. This is due to epitope masking, reduced affinity, or changes in selectivity. A more definitive method for demonstrating protein ubiquitination is to couple immunoprecipitation with digestion by a broad spectrum deubiquitinase (DUB) prior to immunoblot analysis. An increased signal for the protein of interest (POI) after DUB treatment is a clear indication that the protein was ubiquitinated even if there was no clear reactivity in the untreated sample.