Learn More
Aviva Systems Biology Corp UBN1 Antibody - C-terminal region (ARP47638_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47638_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UBN1. Alternate names: VT, VT4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53523