Learn More
Waters Corp μBondapak C18 Column, 125Å, 10 μm, 3.9 mm X 150 mm
Supplier:  Waters Corp WAT086684
Encompass_Preferred
μBondapak C18 columns are general purpose, silica-based, reversed-phase C18 columns utilizing 10 μm particle technology. They serve as a starting point for preparative chromatography, offering a balance of resolution, throughput, and cost. These columns are manufactured under strict quality control processes to ensure high quality and reproducible results.
  • Year-to-year reproducibility is achieved through stringent quality control batch tests.
  • Offers a balance between resolution, throughput, and cost, particularly for preparative chromatography.
  • Manufactured with tightly maintained quality control processes and narrow specification ranges, ensuring high quality and reproducible results across chromatographic applications.
  • Supported for scientists globally, even for methods requiring irregularly shaped particles.
Catalog No. 50-818-612