Learn More
Waters Corp µBondapak C18 Radial-Pak Column, 125Å, 10 µm, 8 mm X 100 mm, 1/pk
Catalog No. 50785946 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
µBondapak C18 columns are general purpose, silica-based, reversed-phase C18 columns that are based on 10 µm particle technology. As a starting point for preparative chromatography, no other column can provide the balance between resolution, throughput and cost.
Catalog No. 50-785-946 Supplier Waters Corp Supplier No. WAT085721
µBondapak C18 columns are general purpose, silica-based, reversed-phase C18 columns that are based on 10 µm particle technology. As a starting point for preparative chromatography, no other column can provide the balance between resolution, throughput and cost.