Learn More
Waters Corp Waters Column, C18, 10 μm, 125 Å, 4.6 mm, 150 mm
Catalog No. 50785570 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
μBondapak C18 columns are general-purpose, silica-based, reversed-phase C18 columns based on 10 μm particle technology. They are designed for preparative chromatography, offering a balance between resolution, throughput, and cost. These columns are manufactured in state-of-the-art facilities, adhering to stringent cGMP and ISO 9001 standards.
  • General-purpose, silica-based, reversed-phase C18 column
  • 10 μm particle technology
  • Designed for preparative chromatography
  • Offers a balance between resolution, throughput, and cost
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Manufactured to stringent cGMP and ISO 9001 standards
Catalog No. 50-785-570 Supplier Waters Corp Supplier No. WAT044370
Add to Cart
Add to Cart
μBondapak C18 columns are general-purpose, silica-based, reversed-phase C18 columns based on 10 μm particle technology. They are designed for preparative chromatography, offering a balance between resolution, throughput, and cost. These columns are manufactured in state-of-the-art facilities, adhering to stringent cGMP and ISO 9001 standards.
  • General-purpose, silica-based, reversed-phase C18 column
  • 10 μm particle technology
  • Designed for preparative chromatography
  • Offers a balance between resolution, throughput, and cost
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Manufactured to stringent cGMP and ISO 9001 standards