Learn More
Waters Corp Waters Guard Cartridge, C18, 10 μm, 3.9 mm X 20 mm
Catalog No. 50785578 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Designed to be used with C18 columns.
  • Protects analytical columns from damaging contaminants.
  • Enhances and improves column analytical performance.
  • Extends the lifetime of analytical columns.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Manufactured under cGMP, ISO 9001 conditions.
  • Each unit is tested and certified for use.
  • Tolerates pressures up to 6000 psi (415 bar).
  • Made using identical packing materials as analytical columns.
  • Features BEH Technology C18.
Catalog No. 50-785-578 Supplier Waters Corp Supplier No. WAT044480
  • Designed to be used with C18 columns.
  • Protects analytical columns from damaging contaminants.
  • Enhances and improves column analytical performance.
  • Extends the lifetime of analytical columns.
  • Maintains UPLC, UHPLC, and HPLC separation efficiency.
  • Manufactured under cGMP, ISO 9001 conditions.
  • Each unit is tested and certified for use.
  • Tolerates pressures up to 6000 psi (415 bar).
  • Made using identical packing materials as analytical columns.
  • Features BEH Technology C18.