Learn More
Waters Corp Waters Column, Cyano (CN), 10 μm, 3.9 mm x 300 mm, 125 Å
Catalog No. 50785903 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Cyano (CN) Column is a versatile, silica-based, reversed-phase column, built with ultra-high purity spherical silica gel and cyanopropyl functional groups. Ideal for analytical and preparative procedures, it excels in irregular particle analysis, balancing resolution, throughput, and cost.
  • General-purpose, silica-based, reversed-phase column
  • Features ultra-high purity spherical silica gel with cyanopropyl functional groups
  • Supports analytical and preparative procedures, including irregular particle analysis
  • Offers excellent stability from maximized bond coverage
  • Provides rapid elution for hydrophobic molecules and unique selectivity for polar compounds
  • Delivers good peak shape for strongly basic compounds
  • Effective for highly hydrophobic compounds not eluted by standard C18/C8 columns
  • Operates up to 6000 psi (415 Bar) maximum pressure
Catalog No. 50-785-903 Supplier Waters Corp Supplier No. WAT084042
This Cyano (CN) Column is a versatile, silica-based, reversed-phase column, built with ultra-high purity spherical silica gel and cyanopropyl functional groups. Ideal for analytical and preparative procedures, it excels in irregular particle analysis, balancing resolution, throughput, and cost.
  • General-purpose, silica-based, reversed-phase column
  • Features ultra-high purity spherical silica gel with cyanopropyl functional groups
  • Supports analytical and preparative procedures, including irregular particle analysis
  • Offers excellent stability from maximized bond coverage
  • Provides rapid elution for hydrophobic molecules and unique selectivity for polar compounds
  • Delivers good peak shape for strongly basic compounds
  • Effective for highly hydrophobic compounds not eluted by standard C18/C8 columns
  • Operates up to 6000 psi (415 Bar) maximum pressure