Learn More
Waters Corp Waters Prep Column, Amino (NH2), 10 μm, 7.8 mm X 300 mm
Catalog No. 50785908 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The μBondapak Amino (NH2) Prep Column is an HPLC column recognized for Waters’ stringent QC batch testing and provides reproducible, high-performance chromatography. It facilitates scaling from preparative to analytical applications due to its method transfer features and offers a wide pH range for separating acids, bases, and neutrals.
  • Designed for methods requiring packings.
  • Undergoes strict quality control testing by scientists.
  • Widely referenced and demonstrates reproducibility for over 40 years.
  • Provides a wide and usable pH range (2 to 8 pH).
  • Can be adapted to the Guard-Pak format using Guard-Pak inserts.
Catalog No. 50-785-908 Supplier Waters Corp Supplier No. WAT084178
The μBondapak Amino (NH2) Prep Column is an HPLC column recognized for Waters’ stringent QC batch testing and provides reproducible, high-performance chromatography. It facilitates scaling from preparative to analytical applications due to its method transfer features and offers a wide pH range for separating acids, bases, and neutrals.
  • Designed for methods requiring packings.
  • Undergoes strict quality control testing by scientists.
  • Widely referenced and demonstrates reproducibility for over 40 years.
  • Provides a wide and usable pH range (2 to 8 pH).
  • Can be adapted to the Guard-Pak format using Guard-Pak inserts.