Learn More
Waters Corp Waters Sentry Guard Cartridge, Amino (NH2), 10 μm, 3.9 mm X 20 mm
Catalog No. 50785630 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sentry Guard Cartridge is designed for methods requiring μBondapak packings, offering reliable performance. It meets stringent quality control standards, ensuring reproducible and high-performance chromatographic results. μBondapak columns are globally recognized and highly referenced.
  • Amino (NH2) chemistry
  • Normal-phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Irregular particle shape
  • 10 μm particle size
  • 125 Å pore size
  • 330 surface area
  • Designed for HPLC systems
  • USP classification L8
  • 3.9 mm inner diameter
  • 20 mm length
  • 4 % carbon load
  • Packaged with 2 units per package
Catalog No. 50-785-630 Supplier Waters Corp Supplier No. WAT046865
This Sentry Guard Cartridge is designed for methods requiring μBondapak packings, offering reliable performance. It meets stringent quality control standards, ensuring reproducible and high-performance chromatographic results. μBondapak columns are globally recognized and highly referenced.
  • Amino (NH2) chemistry
  • Normal-phase separation mode
  • Silica particle substrate
  • pH range of 2 to 8
  • Maximum pressure of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Irregular particle shape
  • 10 μm particle size
  • 125 Å pore size
  • 330 surface area
  • Designed for HPLC systems
  • USP classification L8
  • 3.9 mm inner diameter
  • 20 mm length
  • 4 % carbon load
  • Packaged with 2 units per package