Learn More
Waters Corp Waters μBondapak Guard-Pak Insert, Amino (NH2), 125Å, 10 μm, 10-pack
Catalog No. 50785253 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard-pak insert is designed for methods requiring the use of specific packings, offering reliable performance. It provides reproducible, high-performance chromatographic separation, adhering to stringent quality control standards. This product is widely referenced and requested, highlighting its established presence in chromatographic applications.
  • Amino (NH2) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Operates within a pH range of 2 to 8 pH
  • Withstands maximum pressures of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Irregular particle shape
  • Particle size of 10 μm
  • Pore size of 125 Å
  • Designed in a guard-pak insert format
  • Surface area of 330
  • Compatible with HPLC systems
  • USP Classification L8
  • 4% carbon load
Catalog No. 50-785-253 Supplier Waters Corp Supplier No. WAT026760
This guard-pak insert is designed for methods requiring the use of specific packings, offering reliable performance. It provides reproducible, high-performance chromatographic separation, adhering to stringent quality control standards. This product is widely referenced and requested, highlighting its established presence in chromatographic applications.
  • Amino (NH2) chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • Operates within a pH range of 2 to 8 pH
  • Withstands maximum pressures of 6000 psi (415 Bar)
  • Not endcapped
  • High silanol activity
  • Irregular particle shape
  • Particle size of 10 μm
  • Pore size of 125 Å
  • Designed in a guard-pak insert format
  • Surface area of 330
  • Compatible with HPLC systems
  • USP Classification L8
  • 4% carbon load