Learn More
Waters Corp µBondapak Phenyl Column, 125Å, 10 µm, 3.9 mm X 150 mm, 1/pk
Catalog No. 50785956 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
If your method calls for µBondapak packings, there is only one µBondapak packing material. Many companies claim to have columns with µBondapak-like selectivity but not one has ever passed Waters stringent QC batch test. Waters set the standard for reproducible, high-performance chromatographic columns. Waters µBondapak columns are still the most widely referenced and requested columns in the world.
Catalog No. 50-785-956 Supplier Waters Corp Supplier No. WAT086680
If your method calls for µBondapak packings, there is only one µBondapak packing material. Many companies claim to have columns with µBondapak-like selectivity but not one has ever passed Waters stringent QC batch test. Waters set the standard for reproducible, high-performance chromatographic columns. Waters µBondapak columns are still the most widely referenced and requested columns in the world.