Learn More
Waters Corp Phenyl Guard Cartridge, 10 μm, 125 Å
Catalog No. 50785627 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed to protect analytical columns from contamination, thereby extending their lifespan and improving chromatographic performance. It removes chemical and particle impurities from the mobile phase stream, ensuring consistent separation efficiency across UPLC, UHPLC, and HPLC systems. Manufactured under rigorous cGMP and ISO 9001 conditions, each unit is thoroughly tested and approved to ensure optimal function and enhanced selectivity, sensitivity, and speed of studies.
  • Phenyl chemistry for reversed-phase separation
  • Silica particle substrate with 10 μm irregular particles
  • Pore size of 125 Å
  • Withstands maximum pressure of 6000 psi (415 bar)
  • Designed to protect analytical columns from contamination
  • Manufactured under strict cGMP and ISO 9001 conditions
Catalog No. 50-785-627 Supplier Waters Corp Supplier No. WAT046850
This guard cartridge is designed to protect analytical columns from contamination, thereby extending their lifespan and improving chromatographic performance. It removes chemical and particle impurities from the mobile phase stream, ensuring consistent separation efficiency across UPLC, UHPLC, and HPLC systems. Manufactured under rigorous cGMP and ISO 9001 conditions, each unit is thoroughly tested and approved to ensure optimal function and enhanced selectivity, sensitivity, and speed of studies.
  • Phenyl chemistry for reversed-phase separation
  • Silica particle substrate with 10 μm irregular particles
  • Pore size of 125 Å
  • Withstands maximum pressure of 6000 psi (415 bar)
  • Designed to protect analytical columns from contamination
  • Manufactured under strict cGMP and ISO 9001 conditions