Learn More
Waters Corp Waters μBondapak Phenyl Guard-Pak Insert, 125 Å, 10 μm
Catalog No. 50785250 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The μBondapak Phenyl Guard-Pak Insert enhances lab equipment functionality and extends the lifespan of μBondapak Phenyl Columns by removing chemical and particle pollutants. It features the same packing materials as HPLC columns and cartridges, compatible with 10 μm particle columns, and contains a microparticulate calcium cation-exchange gel. Manufactured under rigorous guidelines, it undergoes stringent quality control for batch-to-batch repeatability and performance.
  • Extends lifespan of μBondapak Phenyl Columns by removing chemical/particle pollutants.
  • Improves analytical lab equipment functionality.
  • Uses same packing materials as Guard-Pak inserts, HPLC columns/cartridges.
  • Manufactured with pure reagents under rigorous guidelines.
  • Stringent QC/chromatographic tests ensure batch repeatability and performance.
  • Each piece individually examined for quality.
Catalog No. 50-785-250 Supplier Waters Corp Supplier No. WAT026745
The μBondapak Phenyl Guard-Pak Insert enhances lab equipment functionality and extends the lifespan of μBondapak Phenyl Columns by removing chemical and particle pollutants. It features the same packing materials as HPLC columns and cartridges, compatible with 10 μm particle columns, and contains a microparticulate calcium cation-exchange gel. Manufactured under rigorous guidelines, it undergoes stringent quality control for batch-to-batch repeatability and performance.
  • Extends lifespan of μBondapak Phenyl Columns by removing chemical/particle pollutants.
  • Improves analytical lab equipment functionality.
  • Uses same packing materials as Guard-Pak inserts, HPLC columns/cartridges.
  • Manufactured with pure reagents under rigorous guidelines.
  • Stringent QC/chromatographic tests ensure batch repeatability and performance.
  • Each piece individually examined for quality.