Learn More
Aviva Systems Biology Corp Ubox5 Antibody - middle region (ARP43174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ubox5. It was validated on Western Blot. . Alternate names: 1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-061