Learn More
Medchemexpress LLC HY-107604 5mg Medchemexpress, UBP 302 CAS:745055-91-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1076045MG
Encompass_Preferred
Medchemexpress, HY-107604 5mg UBP 302 CAS:745055-91-8 UBP 302 is a potent and selective GLUK5-subunit containing kainate receptor antagonist (apparent Kd=402 nM), and displays very little affinity on GluK2 (GluR6) kainate receptors. Anxiolytic effects[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8437