Learn More
Aviva Systems Biology Corp UBP1 Antibody - middle region (ARP33478_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33478_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBP1. Alternate names: LBP1A, LBP1B, LBP-1B, LBP-1aApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55038