Learn More
Ubpbio 6xHis-TEV Protease, 1 MG
Catalog No. 501393686
Encompass_Preferred
Protein Purification Reagents - 6xHis-TEV Protease -Tobacco Etch Virus (TEV) protease is a cysteine protease that is commonly used to remove fusion tags, including His, GST and MBP, at the N or C termini of recombinant proteins. TEV protease specifically recognizes a seven amino acid sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly/Ser (ENLYFQG/S) and cleaves between Gln and Gly/Ser under a wide range of conditions.
Catalog No. 50-139-3686 Supplier Ubpbio Supplier No. P10011000
Protein Purification Reagents - 6xHis-TEV Protease -Tobacco Etch Virus (TEV) protease is a cysteine protease that is commonly used to remove fusion tags, including His, GST and MBP, at the N or C termini of recombinant proteins. TEV protease specifically recognizes a seven amino acid sequence Glu-Asn-Leu-Tyr-Phe-Gln-Gly/Ser (ENLYFQG/S) and cleaves between Gln and Gly/Ser under a wide range of conditions.