Learn More
Ubpbio Agar Powder, 500 GR, CAS# 9002-18-0
Catalog No. 501393673
Encompass_Preferred
Protein Expression Reagents – Agar – Used as a solidifying agent, mainly used for preparing microbiological LB plate at 1.5%.
Catalog No. 50-139-3673 Supplier Ubpbio Supplier No. P1110500
Protein Expression Reagents – Agar – Used as a solidifying agent, mainly used for preparing microbiological LB plate at 1.5%.