Learn More
Ubpbio Agar Powder, 5000 GR, CAS# 9002-18-0
Catalog No. 501393675
Encompass_Preferred
Protein Expression Reagents – Agar – Used as a solidifying agent, mainly used for preparing microbiological LB plate at 1.5%.
Catalog No. 50-139-3675 Supplier Ubpbio Supplier No. P11105000
Protein Expression Reagents – Agar – Used as a solidifying agent, mainly used for preparing microbiological LB plate at 1.5%.