Learn More
Ubpbio GST-HRV 3C Protease, 1 MG
Catalog No. 501393688
Encompass_Preferred
Protein Purification Reagents - GST-HRV 3C Protease -Human rhinovirus 3C protease (HRV 3C Protease) is a cysteine protease that recognizes the Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro sequence (also called the PreScission site), and cleaves between Gln and Gly. The recombinant GST-HRV 3C can be used to cleave the GST tag in fusion proteins expressed using the pGEX-6p vector or other vectors, then removed by glutathione resin. It exhibits high specificity and activity at 4 0C in an overnight incubation reaction containing 1 mass unit of HRR 3C and 100 mass units of targeting protein.
Catalog No. 50-139-3688 Supplier Ubpbio Supplier No. P10021000
Protein Purification Reagents - GST-HRV 3C Protease -Human rhinovirus 3C protease (HRV 3C Protease) is a cysteine protease that recognizes the Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro sequence (also called the PreScission site), and cleaves between Gln and Gly. The recombinant GST-HRV 3C can be used to cleave the GST tag in fusion proteins expressed using the pGEX-6p vector or other vectors, then removed by glutathione resin. It exhibits high specificity and activity at 4 0C in an overnight incubation reaction containing 1 mass unit of HRR 3C and 100 mass units of targeting protein.