Learn More
Ubpbio TCEP Powder, 10 GR, CAS# 51805-45-9
Catalog No. 501393682
Encompass_Preferred
Reducing Agents - TCEP - TCEP (tris(2-carboxyethyl)phosphine) is a strong reducing agent. Compared to dithiothreitol (DTT) and β-mercaptoethanol, TCEP has the advantages of being odorless, a more powerful reducing agent, an irreversible reducing agent, more hydrophilic, and more resistant to oxidation in air. It is soluble in water and stable at neutral pH. It does not reduce metals used in immobilized metal affinity chromatography such as Ni-NTA resin. TCEP is particularly useful when labeling cysteine residues with maleimides.
Catalog No. 50-139-3682 Supplier Ubpbio Supplier No. P102010
Reducing Agents - TCEP - TCEP (tris(2-carboxyethyl)phosphine) is a strong reducing agent. Compared to dithiothreitol (DTT) and β-mercaptoethanol, TCEP has the advantages of being odorless, a more powerful reducing agent, an irreversible reducing agent, more hydrophilic, and more resistant to oxidation in air. It is soluble in water and stable at neutral pH. It does not reduce metals used in immobilized metal affinity chromatography such as Ni-NTA resin. TCEP is particularly useful when labeling cysteine residues with maleimides.