Learn More
Aviva Systems Biology Corp UBQLN2 Antibody - N-terminal region (ARP59353_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59353_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBQLN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human UBQLN2. Alternate names: CHAP1, DSK2, FLJ10167, FLJ56541, HRIHFB2157, N4BP4, PLIC2, ALS15ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-672