Learn More
Sino Biological UBQLN2 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 202831T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human UBQLN2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-6427