Learn More
Aviva Systems Biology Corp UBQLN4 Antibody - middle region (ARP57355_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57355_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBQLN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBQLN4. Alternate names: A1U, C1orf6, UBIN, CIP75ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53854