Learn More
ABclonal Technology UBQLN4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15869500UL
Encompass_Preferred
Enables K48-linked polyubiquitin modification-dependent protein binding activity and identical protein binding activity. Involved in cellular response to DNA damage stimulus, negative regulation of double-strand break repair via homologous recombination, and regulation of cellular catabolic process. Located in several cellular components, including autophagosome, nucleoplasm, and site of DNA damage. Part of protein-containing complex. Colocalizes with cytosolic proteasome complex and nuclear proteasome complex.
Catalog No. 50-001-05695