Learn More
Aviva Systems Biology Corp UBQLNL Antibody - middle region (ARP52793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBQLNL. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBQLNL. Alternate names: MGC20470, MGC26958ApplicationWB. Species reactivity: Human
Catalog No. 50-285-766