Learn More
Aviva Systems Biology Corp UBR2 Antibody - C-terminal region (ARP43188_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43188_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human UBR2. Alternate names: C6orf133, DKFZp686C08114, KIAA0349, MGC71112, RP3-392M17.3, bA49A4.1, dJ242G1.1, dJ392M17.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58458