Learn More
Aviva Systems Biology Corp UBTF Antibody - middle region (ARP33480_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33480_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBTF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human UBTF. Alternate names: NOR-90, UBF, UBF1, UBF-1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55039