Learn More
Aviva Systems Biology Corp Ubxn2a Antibody - middle region (ARP53408_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53408_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ubxn2a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Ubxd4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-836