Learn More
Aviva Systems Biology Corp UBXN6 Antibody - C-terminal region (ARP59293_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59293_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UBXN6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human UBXN6. Alternate names: DKFZp667D109, FLJ00394, UBXD1, UBXDC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-646