Learn More
Medchemexpress LLC UC-112 | 383392-66-3 | 98.9% | 100 MG
SDP
Catalog No. 5000341129
Encompass_Preferred
UC-112 is a survivin and XIAP modulator exhibiting anticancer activity. It functions by selectively downregulating and degrading survivin through the ubiquitin-mediated proteasomal pathway. This compound also effectively reduces XIAP levels in in vivo tumor models, leading to the activation of caspase-3/7 and caspase-9, and ultimately inducing apoptosis in cancer cells. It is suitable for research on melanoma, prostate cancer, and other cancer-related studies.
  • Selectively downregulates and degrades survivin
  • Reduces XIAP levels in in vivo tumor models
  • Activates caspase-3/7 and caspase-9
  • Induces cancer cell apoptosis
  • Inhibits proliferation of A375, M14, PC-3, and DU145 cancer cells
  • Alters cell cycle distribution in A375 melanoma cells
  • Inhibits human melanoma tumor growth in nude mice
Catalog No. 50-003-41129 Supplier Medchemexpress LLC Supplier No. HY12842100MG
UC-112 is a survivin and XIAP modulator exhibiting anticancer activity. It functions by selectively downregulating and degrading survivin through the ubiquitin-mediated proteasomal pathway. This compound also effectively reduces XIAP levels in in vivo tumor models, leading to the activation of caspase-3/7 and caspase-9, and ultimately inducing apoptosis in cancer cells. It is suitable for research on melanoma, prostate cancer, and other cancer-related studies.
  • Selectively downregulates and degrades survivin
  • Reduces XIAP levels in in vivo tumor models
  • Activates caspase-3/7 and caspase-9
  • Induces cancer cell apoptosis
  • Inhibits proliferation of A375, M14, PC-3, and DU145 cancer cells
  • Alters cell cycle distribution in A375 melanoma cells
  • Inhibits human melanoma tumor growth in nude mice