Learn More
Azzota Corp UC 30L 35kHz 600W 20-69C
Catalog No. 5000887259
Encompass_Preferred
Azzota Low-frequency 35KHZ Fixed Power Ultrasonic Cleaners Benchtop 30L 600W 20 C-69 C 500x300x200mm 1-99 min with drain
Catalog No. 50-008-87259 Supplier Azzota Corp Supplier No. BLF600T
Azzota Low-frequency 35KHZ Fixed Power Ultrasonic Cleaners Benchtop 30L 600W 20 C-69 C 500x300x200mm 1-99 min with drain