Learn More
Santa Cruz Biotechnology UltraCruz Cryo-marker Dual point, Green, 10/pk
Catalog No. NC1512917
Encompass
Cryo-marker is a cryogenic waterproof and permanent marker used for indelibly marking a wide variety of surfaces in the laboratory, including teflon.
Catalog No. NC1512917 Supplier Santa Cruz Biotechnology Supplier No. SC360979
Cryo-marker is a cryogenic waterproof and permanent marker used for indelibly marking a wide variety of surfaces in the laboratory, including teflon.