Learn More
Yamato Scientific America, Inc. Yamato Undercounter & Countertop Refrigerator 1ºc To 7ºc, 4.6 Cu.Ft. With Glass Door, Cycle Defrost, 115V
Supplier:  Yamato Scientific America, Inc. UCR101G
Encompass_Preferred
Undercounter & Benchtop lab and pharmacy refrigerators are designed to provide safe sample storage at an average temperature of +4°C.
Catalog No. NC1869712