Learn More
Medchemexpress LLC HY-112780 100mg , UC2288 CAS:1394011-91-6 Purity:>98%
SDP
Catalog No. 502260557
Encompass_Preferred
HY-112780 100mg UC2288 CAS: 1394011-91-6 UC2288 is a novel, cell-permeable, and orally active p21 attenuator (relatively selective activity for p21), which is synthesized based Sorafenib (HY-10201). UC2288 decreases p21 mRNA expression independently of p53, and attenuates p21 protein levels with minimal effect on p21 protein stability. UC2288 has no inhibition of VEGFR2 and Raf kinases even at 10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0557 Supplier Medchemexpress LLC Supplier No. HY112780100MG
HY-112780 100mg UC2288 CAS: 1394011-91-6 UC2288 is a novel, cell-permeable, and orally active p21 attenuator (relatively selective activity for p21), which is synthesized based Sorafenib (HY-10201). UC2288 decreases p21 mRNA expression independently of p53, and attenuates p21 protein levels with minimal effect on p21 protein stability. UC2288 has no inhibition of VEGFR2 and Raf kinases even at 10 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.