Learn More
Abcam UC2288, p21 inhibitor, 5MG
Catalog No. 502583214
Encompass_Preferred
MW 481.8 Da, Purity >98%. Cell-permeable, phenylcyclohexyl-urea based compound that selectively downregulates the expression of p21, independent of p53 expression, at either transcription or post-transcriptional level. Attenuates p21 protein levels with minimal effect on p21 protein stability. Has no significant effect on the activities of Raf kinases, VEGFR2 kinase, or the phosphorylation state of ERK. Effectively inhibits the growth of multiple cancer cell lines (GI₅₀ = ~ 10 μM against NCI60 cell lines).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3214 Supplier Abcam Supplier No. AB1469695MG
MW 481.8 Da, Purity >98%. Cell-permeable, phenylcyclohexyl-urea based compound that selectively downregulates the expression of p21, independent of p53 expression, at either transcription or post-transcriptional level. Attenuates p21 protein levels with minimal effect on p21 protein stability. Has no significant effect on the activities of Raf kinases, VEGFR2 kinase, or the phosphorylation state of ERK. Effectively inhibits the growth of multiple cancer cell lines (GI₅₀ = ~ 10 μM against NCI60 cell lines).
The product is subject to the following: Abcam Restricted Use Statement