Learn More
Medchemexpress LLC HY-133122 10mg , UCB-9260 CAS:1515888-53-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13312210MG
Encompass_Preferred
HY-133122 10mg UCB-9260 CAS: 1515888-53-5 UCB-9260, an orally active compound, inhibits TNF signaling by stabilising an asymmetric form of the trimer. UCB-9260 is selective for TNF over other superfamily members, and binds TNF with a similar Kd of 13 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9696