Learn More
Medchemexpress LLC HY-112613 10mg Medchemexpress, UCB9608 CAS:1616413-96-7 Purity:>98%
SDP
Catalog No. 501969117
Encompass_Preferred
Medchemexpress, HY-112613 10mg UCB9608 CAS:1616413-96-7 UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9117 Supplier Medchemexpress LLC Supplier No. HY11261310MG
Medchemexpress, HY-112613 10mg UCB9608 CAS:1616413-96-7 UCB9608 is a potent, selective and orally active PI4KIIIβ inhibitor, with an IC50 of 11 nM, selective over PI3KC2 α, β, and γ lipid kinases. UCB9608 improves metabolic stability and exhibits excellent pharmacokinetic profile, acts as a potent immunosuppressive agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.