Learn More
Medchemexpress LLC HY-125959 5mg Medchemexpress, Ucf-101 CAS:313649-08-0 Purity:>98%
SDP
Catalog No. 502081088
Encompass_Preferred
Medchemexpress, HY-125959 5mg Ucf-101 CAS:313649-08-0 Ucf-101 is a selctive and competitive inhibitor of pro-apoptotic protease Omi/HtrA2, with an IC50 of 9.5 μM for His-Omi. Ucf-101 exhibits very little activity against various other serine proteases (IC50>200 μM). Ucf-101 has a natural red fluorescence at 543 nm that is used to monitor its ability to enter mammalian cells. Ucf-101 has a significant cardioprotective effect against MI/R injury and also has certain neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1088 Supplier Medchemexpress LLC Supplier No. HY1259595MG
Medchemexpress, HY-125959 5mg Ucf-101 CAS:313649-08-0 Ucf-101 is a selctive and competitive inhibitor of pro-apoptotic protease Omi/HtrA2, with an IC50 of 9.5 μM for His-Omi. Ucf-101 exhibits very little activity against various other serine proteases (IC50>200 μM). Ucf-101 has a natural red fluorescence at 543 nm that is used to monitor its ability to enter mammalian cells. Ucf-101 has a significant cardioprotective effect against MI/R injury and also has certain neuroprotective effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.