Learn More
Enzo Life Sciences UCH-L3 (human), (recombinant) (His-tag) (50 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW97450050
Encompass_Preferred
Alternative name: Ubiquitin C terminal hydrolase L3. Purity: ≥90% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS, pH 7.6, containing 1mM dithiothreitol (DTT). Source: Produced in E. coli. Long Term Storage: -80°C.
Catalog No. 50-201-2327