Learn More
Aviva Systems Biology Corp UCHL5 Antibody - C-terminal region (ARP59295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UCHL5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human UCHL5. Alternate names: CGI-70, INO80R, UCH37, UCH-L5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-648